o-Xylen, Supelco®
Leverantör: Merck
Fara
Synonyms: 1,2-Dimethylbenzene
M-Clarity™ quality level = MQ100
95660-5ML 95660-50ML
95660-5MLEA 1010 SEK
95660-5ML 95660-50ML
o-Xylen, Supelco®
o-Xylen
Formel: C₈H₁₀
MW: 106,17 g/mol
Kokpunkt: 144 °C (1013 hPa)
Smältpunkt: –25 °C
Densitet: 0,88 g/cm³ (20 °C)
Flampunkt: 30 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00008519
CAS nummer: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=950bdbbeadd692c67ac79579692ca8939279051e&vid=131925d29082c5a5cd4eb302adcafab48945365b&prid=web&clid=SAPDAM