3,5-Dimethylthiophenol

Leverantör: Thermo Scientific
Varning

Synonyms: 1,3-Dimethyl-5-mercaptobenzene, 3,5-Dimethylbenzene-1-thiol, 3,5-Dimethylbenzenethiol, 5-Mercapto-m-xylene

SB01213DA SB01213EA
MAYBSB01213DAEA 0 SEK
MAYBSB01213DA MAYBSB01213EA
3,5-Dimethylthiophenol
3,5-Dimethylthiophenol
Formel: C₈H₁₀S
MW: 138.23 g/mol
Kokpunkt: 96
Densitet: 1.02 g/cm³ (20 °C)
MDL Number: MFCD00040226
CAS nummer: 38360-81-5
EINECS: 253-901-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR