3,4-Dimethylthiophenol ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2-Dimethyl-4-mercaptobenzene, 4-Mercapto-o-xylene, 3,4-Dimethylbenzene-1-thiol, 3,4-Dimethylbenzenethiol
B23168.03 B23168.06 B23168.14
B23168.03EA 1250 SEK
B23168.03 B23168.06 B23168.14
3,4-Dimethylthiophenol ≥98%
3,4-Dimethylthiophenol
Purity: 98% CAS No.: 18800-53-8 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 90°(194°F) Refractive Index: 1.5736 Odour: Unpleasant Boiling Point: 218° Beilstein Reference: 2040353 UN No.: 2810 MDL No.: MFCD00010023
Formel: C₈H₁₀S
MW: 138,23 g/mol
Kokpunkt: 218 °C
Smältpunkt: 54…56 °C
Densitet: 1,027
Flampunkt: 90 °C (194 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010023
CAS nummer: 18800-53-8
EINECS: 242-587-8
UN: 2810
ADR: 6.1,III