2,5-Xylenol zS, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: 2,5-Dimetylfenol
MQ200
8.03771.0005 8.03771.0250
8.03771.0005EA 286 SEK
8.03771.0005 8.03771.0250
2,5-Xylenol zS, Sigma-Aldrich®
2,5-Xylenol
Formel: C₈H₁₀O
MW: 122.17 g/mol
Kokpunkt: 212
Smältpunkt: 75
Densitet: 1.169 g/cm³ (15 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002237
CAS nummer: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1 II
Merck Index: 12,10215
Order Now
Specifikation testresultat
Solubility (20 °C) slightly soluble
Melting Point 75 °C
Molar Mass 122.16 g/mol
Boiling Point 211 °C
Vapor Pressure 0.208 hPa (25 °C)
Flash Point 80 - 95 °C
Explosion Limit 1.4 %(V)
Density 0.9 g/cm3 (75 °C)
Synonyms 2,5-DMP, p-Xylenol
Hill Formula C8H10O
Chemical Formula 2,5-(CH3)2C6H3OH