2,6-Dimethylanisole ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B20365.14 B20365.22
B20365.14EA 1500 SEK
B20365.14 B20365.22
2,6-Dimethylanisole ≥98%
2,6-Dimethylanisole
Purity: 98+% CAS No.: 1004-66-6 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 67°(152°F) Refractive Index: 1.5030 Boiling Point: 181-182° Beilstein Reference: 1858509 MDL No.: MFCD00008380
Formel: C₉H₁₂O
MW: 136.19 g/mol
Kokpunkt: 181…182 °C
Densitet: 0.962
Flampunkt: 67 °C (152 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008380
CAS nummer: 1004-66-6
EINECS: 213-723-3