2-Fluoro-m-xylene ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene
A10424.06 A10424.14
A10424.06EA 1475 SEK
A10424.06 A10424.14
2-Fluoro-m-xylene ≥98%
2-Fluoro-m-xylene
Purity: 98% CAS No.: 443-88-9 Molecular Formula: C8H9F Formula Weight: 124.16 Flash Point: 30°(86°F) Refractive Index: 1.4790 Odour: Aromatic Boiling Point: 147-148° Beilstein Reference: 1857413 UN No.: 1993 MDL No.: MFCD00039215
Formel: C₈H₉F
MW: 124,16 g/mol
Kokpunkt: 147…148 °C
Densitet: 0,988
Flampunkt: 30 °C (86 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039215
CAS nummer: 443-88-9
EINECS: 207-144-5
UN: 1993
ADR: 3,III