4-tert-Butyl-α-chlorotoluene 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: p-tert-Butylbenzyl chloride, 4-tert-Butylbenzyl chloride, p-tert-Butyl-α-chlorotoluene, 1-(tert-Butyl)-4-(chloromethyl)benzene
345490050 345490250
ACRO345490050EA 640 SEK
ACRO345490050 ACRO345490250
4-tert-Butyl-α-chlorotoluene 97%
4-tert-Butyl-α-chlorotoluene
Formel: C₁₁H₁₅Cl
MW: 182.69 g/mol
Kokpunkt: 98…103 °C (7 mmHg)
Densitet: 0.94 g/cm³
Flampunkt: 94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000918
CAS nummer: 19692-45-6
EINECS: 243-228-8
UN: 1760
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5205 to 1.5225 (20°C, 589 nm)
GC ≥96.0 %