4-Nitrophenethyl bromide ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene

N0591-25G N0591-100G
TCIAN0591-25GEA 1025 SEK
TCIAN0591-25G TCIAN0591-100G
4-Nitrophenethyl bromide ≥98.0%
4-Nitrophenethyl bromide
Formel: C₈H₈BrNO₂
MDL Number: MFCD00007386
CAS nummer: 5339-26-4
EINECS: 226-271-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d39e6ea864d9b9585175a5d39f4b686140c2aba&vid=7d39e6ea864d9b9585175a5d39f4b686140c2aba&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR