ε-Kaprolaktam ≥99.0%
Leverantör: TCI
|
Varning
|
C0203-25G
C0203-500G
TCIAC0203-25GEA
215
SEK
TCIAC0203-25G
TCIAC0203-500G
ε-Kaprolaktam ≥99.0%
ε-Kaprolaktam
Formel:
C₆H₁₁NO Kokpunkt: 267°C |
MDL Number:
MFCD00006936 CAS nummer: 105-60-2 EINECS: 203-313-2 Merck Index: 14,01761 |
|
Specifikation testresultat
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 69.0 to 72.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...