1-Propyl-2-urea

Leverantör: Thermo Scientific

Synonyms: n-Propylurea, 1-Propylurea

TL00479DA TL00479EA
MAYBTL00479DAEA 0 SEK
MAYBTL00479DA MAYBTL00479EA
1-Propyl-2-urea
1-Propyl-2-urea
Formel: C₄H₁₀N₂O
MW: 102.14 g/mol
MDL Number: MFCD00014793
CAS nummer: 627-06-5
EINECS: 210-981-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR