(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (R)-(+)-1-(4-Chlorophenyl)ethylamine, (R)-1-(4-Chlorophenyl)ethanamine, (R)-1-(4-Chlorophenyl)ethylamine
L19119.03 L19119.06
L19119.03EA 955 SEK
L19119.03 L19119.06
(R)-(+)-4-Chloro-α-methylbenzylamine 97%, ee 98%, ChiPros®
(R)-(+)-4-Chloro-α-methylbenzylamine
Purity: 97% CAS No.: 27298-99-3 Molecular Formula: C8H10ClN Formula Weight: 155.63 Flash Point: 115°(239°F) Refractive Index: 1.5418 Boiling Point: 232° Beilstein Reference: 2412317 Optical Rotation: +31° (neat) UN No.: 2922 MDL No.: MFCD00671639
Formel: C₈H₁₀ClN
MW: 155,63 g/mol
Kokpunkt: 232 °C
Densitet: 1,110
Flampunkt: 115 °C (239 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00671639
CAS nummer: 27298-99-3
UN: 2810
ADR: 6.1,II