(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (S)-1-(3-methoxyphenyl)ethan-1-amine, (S)-(-)-1-(3-Methoxyphenyl)ethylamine
L16324.06 L16324.14 L16324.03
L16324.06EA 2275 SEK
L16324.06 L16324.14 L16324.03
(S)-(-)-3-Methoxy-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-3-Methoxy-α-methylbenzylamine
Purity: 99+% CAS No.: 82796-69-8 Molecular Formula: C9H13NO Formula Weight: 151.21 Flash Point: 100°(212°F) Refractive Index: 1.5335 Melting Point: 5° Boiling Point: 66°/0.38mm Optical Rotation: -27° (neat) UN No.: 2735 MDL No.: MFCD00671656
Formel: C₉H₁₃NO
MW: 151,21 g/mol
Kokpunkt: 66 °C (0,38 mmHg)
Smältpunkt: 5 °C
Densitet: 1,023
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00671656
CAS nummer: 82796-69-8
UN: 2735
ADR: 8,III