(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
Leverantör: Thermo Scientific
Fara
Synonyms: (R)-1-Phenylpropan-1-amine, (R)-(+)-1-Phenylpropylamine
L16319.06 L16319.14 L16319.03
L16319.06EA 1550 SEK
L16319.06 L16319.14 L16319.03
(R)-(+)-α-Ethylbenzylamine ≥99%, ee 98%, ChiPros®
(R)-(+)-α-Ethylbenzylamine
Purity: 99+% CAS No.: 3082-64-2 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 77°(170°F) Refractive Index: 1.5200 Odour: Amine-like Melting Point: -69° Boiling Point: 205° Beilstein Reference: 3029904 Optical Rotation: +20° (neat) UN No.: 2735 MDL No.: MFCD00083057
Formel: C₉H₁₃N
MW: 135,21 g/mol
Kokpunkt: 205 °C
Smältpunkt: –69 °C
Densitet: 0,930
Flampunkt: 77 °C (170 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00083057
CAS nummer: 3082-64-2
UN: 2735
ADR: 8,III