tert-Butylnitrit tech. 90%, Technical Grade
Leverantör: Thermo Scientific
Fara
L16135.14 L16135.22
L16135.14EA 960 SEK
L16135.14 L16135.22
tert-Butylnitrit tech. 90%, Technical Grade
tert-Butylnitrit
Purity: 90% CAS No.: 540-80-7 Molecular Formula: (CH3)3CONO Formula Weight: 103.12 Storage Temperature: 2°C to 8°C Flash Point: -23°(-9°F) Refractive Index: 1.3690 Odour: Sweet Boiling Point: 61-63° Beilstein Reference: 1209339 Merck Reference: 14,1583 UN No.: 2351 MDL No.: MFCD00002055
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokpunkt: 61…63 °C
Densitet: 0,867
Flampunkt: –23 °C (–9 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00002055
CAS nummer: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3,II
Merck Index: 14,1583