(S,S)-(-)-Hydrobenzoin ≥98%
Leverantör: Thermo Scientific
Synonyms: (1S,2S)-1,2-Diphenylethane-1,2-diol
L14082.06 L14082.14
L14082.06EA 2625 SEK
L14082.06 L14082.14
(S,S)-(-)-Hydrobenzoin ≥98%
(S,S)-(-)-Hydrobenzoin
Formel: C₁₄H₁₄O₂
MW: 214,26 g/mol
Smältpunkt: 148…150 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064255
CAS nummer: 2325-10-2
Merck Index: 14,4777