Hydrogen peroxide-urea ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Urea hydrogen peroxide
L13940.14 L13940.22 L13940.36
L13940.14EA 545 SEK
L13940.14 L13940.22 L13940.36
Hydrogen peroxide-urea ≥97%
Hydrogen peroxide-urea
Purity: 97% CAS No.: 124-43-6 Molecular Formula: CH4N2O.H2O2 Formula Weight: 94.07 Odour: Odourless Melting Point: ca 90° dec. Beilstein Reference: 3680414 Merck Reference: 14,1782 UN No.: 1511 MDL No.: MFCD00013119
Formel: CH₆N₂O₃
MW: 94,07 g/mol
Smältpunkt: ∼ 90 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00013119
CAS nummer: 124-43-6
EINECS: 204-701-4
UN: 1511
ADR: 5.1,III
Merck Index: 14,1782