3'-Benzyloxyacetophenone ≥97%
Leverantör: Thermo Scientific
Synonyms:
1-(3-(Benzyloxy)phenyl)ethanone, 1-[3-(Benzyloxy)phenyl]ethan-1-one
B25084.06
B25084.14
B25084.22
B25084.06EA
765
SEK
B25084.06
B25084.14
B25084.22
3'-Benzyloxyacetophenone ≥97%
3'-Benzyloxyacetophenone
Purity: 97%
CAS No.: 34068-01-4
Molecular Formula: C15H14O2
Formula Weight: 226.28
MDL No.: MFCD00026221
Formel:
C₁₅H₁₄O₂ MW: 226,27 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00026221 CAS nummer: 34068-01-4 EINECS: 251-816-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...