Farnesol (mixture of isomers) 96%
Leverantör: Thermo Fisher Scientific
Varning
A19316.06 A19316.14 A19316.22
A19316.06EA 505 SEK
A19316.06 A19316.14 A19316.22
Farnesol (mixture of isomers) 96%
Farnesol (mixture of isomers)
Purity: 96% CAS No.: 4602-84-0 Molecular Formula: C15H26O Formula Weight: 222.37 Flash Point: 146°(295°F) Refractive Index: 1.4890 Odour: Pleasant Boiling Point: 159-160°/10mm Beilstein Reference: 1763926 Merck Reference: 14,3937 MDL No.: MFCD00002918
Formel: C₁₅H₂₆O
Kokpunkt: 159…160 °C (10 mmHg)
Densitet: 0,887
Flampunkt: 146 °C (295 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002918
CAS nummer: 4602-84-0
EINECS: 225-004-1
Merck Index: 14,3937