Galactitol ≥97%
Leverantör: Thermo Scientific
Synonyms: Dulcitol
A18402.14 A18402.22 A18402.36
A18402.14EA 565 SEK
A18402.14 A18402.22 A18402.36
Galactitol ≥97%
Galactitol
Purity: 97% CAS No.: 608-66-2 Molecular Formula: C6H14O6 Formula Weight: 182.17 Melting Point: 187-191° Boiling Point: 275-280°/1mm Merck Reference: 14,4332 MDL No.: MFCD00064288
Formel: HOH₂C(CH(OH))₄CH₂OH
MW: 182,17 g/mol
Kokpunkt: 275…280 °C (1 mmHg)
Smältpunkt: 187…191 °C
Densitet: 1,47
Storage Temperature: Omgivande
MDL Number: MFCD00004708
CAS nummer: 608-66-2
EINECS: 210-165-2
Merck Index: 14,4332