4'-Benzyloxyacetophenone ≥98%

Leverantör: Thermo Scientific

Synonyms: 4-Benzyloxyacetophenone

A18302.06 A18302.14
A18302.06EA 1350 SEK
A18302.06 A18302.14
4'-Benzyloxyacetophenone ≥98%
4'-Benzyloxyacetophenone

Purity: 98% CAS No.: 54696-05-8 Molecular Formula: C15H14O2 Formula Weight: 226.28 Melting Point: 91-92° Beilstein Reference: 1912124

Formel: C₁₅H₁₄O₂
MW: 226,27 g/mol
Smältpunkt: 91…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00017247
CAS nummer: 54696-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR