Perfluorodecahydronaphthalene (mixture of cis and trans isomers) 95%
Leverantör: Thermo Scientific
Synonyms: Perfluoro(decahydronaphthalene), Perfluorodecalin, Perfluorodecahydronaphthalene
A18288.14 A18288.22 A18288.36
A18288.14EA 1150 SEK
A18288.14 A18288.22 A18288.36
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) 95%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Purity: 95% CAS No.: 306-94-5 Molecular Formula: C10F18 Formula Weight: 462.10 Refractive Index: 1.3145 Melting Point: -10 to -7° Boiling Point: 141-142° Beilstein Reference: 2067113 Merck Reference: 14,4183 MDL No.: MFCD00010626
Formel: C₁₀F₁₈
MW: 462,08 g/mol
Kokpunkt: 141…142 °C
Smältpunkt: –10…–7 °C
Densitet: 1,940
Storage Temperature: Omgivande
MDL Number: MFCD00010626
CAS nummer: 306-94-5
EINECS: 206-192-4
Merck Index: 14,4183