1,2-Cyclohexanedione dioxime ≥97%
Leverantör: Thermo Scientific
A16788.06 A16788.14 A16788.22
A16788.06EA 1750 SEK
A16788.06 A16788.14 A16788.22
1,2-Cyclohexanedione dioxime ≥97%
1,2-Cyclohexanedione dioxime
Purity: 97% CAS No.: 492-99-9 Molecular Formula: C6H10N2O2 Formula Weight: 142.16 Melting Point: 181-188° dec. Beilstein Reference: 1909047 Merck Reference: 14,6560 MDL No.: MFCD00001663
Formel: C₆H₁₀N₂O₂
MW: 142,16 g/mol
Smältpunkt: 181…188 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00001663
CAS nummer: 492-99-9
EINECS: 207-769-3
Merck Index: 14,6560