Benzyltrimethylammonium hydroxide 40% (w/w) i metanol
Leverantör: Thermo Scientific
Fara
Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
A14927.0F A14927.AE A14927.AP
A14927.0FEA 5400 SEK
A14927.0F A14927.AE A14927.AP
Benzyltrimethylammonium hydroxide 40% (w/w) i metanol
Benzyltrimethylammonium hydroxide
CAS No.: 100-85-6 Molecular Formula: C10H17NO Formula Weight: 167.25 Flash Point: 15°(59°F) Refractive Index: 1.4270 Odour: Ammonia-like Boiling Point: 65° Beilstein Reference: 3917256 UN No.: 2924 MDL No.: MFCD00008281
Formel: C₁₀H₁₇NO
MW: 167,25 g/mol
Kokpunkt: 65 °C
Densitet: 0,920
Flampunkt: 15 °C (59 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008281
CAS nummer: 100-85-6
EINECS: 202-895-5
UN: 2924
ADR: 3,II