Tetrabutylammonium fluoride 1 M i THF
Leverantör: Thermo Fisher Scientific
Fara
A10588.AE A10588.AC A10588.AP
A10588.AEEA 2775 SEK
A10588.AE A10588.AC A10588.AP
Tetrabutylammonium fluoride 1 M i THF
Tetrabutylammonium fluoride
CAS No.: 429-41-4 Molecular Formula: C16H36FN Formula Weight: 261.47 Flash Point: -17°(1°F) Beilstein Reference: 3570522 UN No.: 2924 MDL No.: MFCD00011747
Formel: C₁₆H₃₆FN
MW: 261.47 g/mol
Densitet: 0.903
Flampunkt: –17 °C (1 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011747
CAS nummer: 429-41-4
EINECS: 207-057-2
UN: 2924
ADR: 3 II