Tetrabutylammonium sulphate 50% (w/w) i vattenlösning
Leverantör: Thermo Scientific
Varning
41723.AE 41723.K4 41723.AP
41723.AEEA 1325 SEK
41723.AE 41723.K4 41723.AP
Tetrabutylammonium sulphate 50% (w/w) i vattenlösning
Tetrabutylammonium sulphate
CAS No.: 2472-88-0 Molecular Formula: [[CH3(CH2)3]4N]2SO4 Formula Weight: 580.06 Physical Form: Liquid Odour: Unpleasant MDL No.: MFCD00216632
Phase transfer catalyst, neutral pH
Formel: C₃₂H₇₂N₂O₄S
MW: 581 g/mol
Kokpunkt: 84 °C
Densitet: 1,01
Storage Temperature: Omgivande
MDL Number: MFCD00216632
CAS nummer: 2472-88-0
EINECS: 219-593-4