2-Bromophenethylamine 99%

Leverantör: Thermo Scientific
Fara

Synonyms: 2-(2-Bromophenyl)ethylamine

350570050 350570200
ACRO350570050EA 2675 SEK
ACRO350570050 ACRO350570200
2-Bromophenethylamine 99%
2-Bromophenethylamine
Formel: C₈H₁₀BrN
MW: 200,08 g/mol
Densitet: 1,396 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD01529874
CAS nummer: 65185-58-2
UN: 2735
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5750 to 1.5770 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR