Tetramethylammonium hydrogen sulphate 99+% för HPLC
Leverantör: Thermo Scientific
Varning
Synonyms: Tetramethylammonium hydrogensulphate, Tetramethylammonium bisulphate
343830050 343830500
ACRO343830050EA 2575 SEK
ACRO343830050 ACRO343830500
Tetramethylammonium hydrogen sulphate 99+% för HPLC
Tetramethylammonium hydrogen sulphate
Formel: C₄H₁₃NO₄S
MW: 171,22 g/mol
Smältpunkt: 295…297 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036149
CAS nummer: 80526-82-5
EINECS: 279-490-5
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV (0.005 M in water)
Titration with NaOH ≥99.0 % (On dry substance)