Ethanolamine hydrochloride 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride
217042500 217040010
ACRO217042500EA 1175 SEK
ACRO217042500 ACRO217040010
Ethanolamine hydrochloride 99+%
Ethanolamine hydrochloride
Formel: C₂H₈ClNO
MW: 97,54 g/mol
Smältpunkt: 84…86 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012892
CAS nummer: 2002-24-6
EINECS: 217-900-6
Order Now
Specifikation testresultat
Appearance White to light beige Crystals and/or agglomerates
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %