4-Methylmorpholine N-oxide hydrate 97%
Leverantör: Thermo Fisher Scientific
Varning
208340250 208341000
ACRO208340250EA 1250 SEK
ACRO208340250 ACRO208341000
4-Methylmorpholine N-oxide hydrate 97%
4-Methylmorpholine N-oxide hydrate
Formel: C₅H₁₁NO₂·1H₂O
Smältpunkt: 70…75 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005947
CAS nummer: 70187-32-5
EINECS: 231-391-8
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤15 % (K.F.)
Titration with HClO4 ≥96.0 % (on dry substance)