Tetrabutylammonium fluoride 1 M i THF
Leverantör: Thermo Scientific
Fara
201950050 201951000 201955000 201950025
ACRO201950050EA 550 SEK
ACRO201950050 ACRO201951000 ACRO201955000 ACRO201950025
Tetrabutylammonium fluoride 1 M i THF
Tetrabutylammonium fluoride
Formel: C₁₆H₃₆FN
MW: 261,47 g/mol
Densitet: 0,887 g/cm³
Flampunkt: –17 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011747
CAS nummer: 429-41-4
EINECS: 207-057-2
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear yellow to amber to light green to brown Solution
Water ≤8 %
Titration with HClO4 0.90 to 1.10 M