2-Methoxyphenethylamine 98%
Leverantör: Thermo Fisher Scientific
Fara
182170250 182171000 182170050
ACRO182170250EA 1900 SEK
ACRO182170250 ACRO182171000 ACRO182170050
2-Methoxyphenethylamine 98%
2-Methoxyphenethylamine
Formel: C₉H₁₃NO
MW: 151.21 g/mol
Kokpunkt: 236…237 °C
Densitet: 1.03 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008186
CAS nummer: 2045-79-6
EINECS: 218-066-6
UN: 2735
ADR: 8 II
Order Now
Specifikation testresultat
Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %