ε-Kaprolaktam 99+%
Leverantör: Thermo Fisher Scientific
Varning
108230010 108230025 108231000 108232500
ACRO108230010EA 860 SEK
ACRO108230010 ACRO108230025 ACRO108231000 ACRO108232500
ε-Kaprolaktam 99+%
ε-Kaprolaktam
Formel: C₆H₁₁NO
Kokpunkt: 268 °C (760 mmHg)
Smältpunkt: 68…71 °C
Flampunkt: 152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006936
CAS nummer: 105-60-2
EINECS: 203-313-2
Order Now
Specifikation testresultat
Appearance White Crystals or flakes
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥99.0 %