Benzyltrimethylammonium hydroxide 40% (w/w) i metanol
Leverantör: Thermo Scientific
Fara
Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
106200010 106202500 106200051
ACRO106200010EA 2175 SEK
ACRO106200010 ACRO106202500 ACRO106200051
Benzyltrimethylammonium hydroxide 40% (w/w) i metanol
Benzyltrimethylammonium hydroxide
Formel: C₁₀H₁₇NO
MW: 167,25 g/mol
Densitet: 0,925 g/cm³
Flampunkt: 15 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008281
CAS nummer: 100-85-6
EINECS: 202-895-5
UN: 3286
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Solution
Infrared spectrum Conforms
Titration with HCl ≥40 %
Titration with HClO4 ≥94 %