2-Bromo-p-xylene ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene
A14358.14 A14358.22
A14358.14EA 845 SEK
A14358.14 A14358.22
2-Bromo-p-xylene ≥98%
2-Bromo-p-xylene
Purity: 98+% CAS No.: 553-94-6 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 79°(174°F) Refractive Index: 1.5510 Melting Point: 9-10° Boiling Point: 199-201° Beilstein Reference: 2040350 MDL No.: MFCD00000074
Formel: C₈H₉Br
MW: 185,06 g/mol
Kokpunkt: 199…201 °C
Smältpunkt: 9…10 °C
Densitet: 1,347
Flampunkt: 79 °C (174 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000074
CAS nummer: 553-94-6
EINECS: 209-055-7