3-Bromo-o-xylene ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-2,3-dimethylbenzene
A15889.06 A15889.14 A15889.22
A15889.06EA 805 SEK
A15889.06 A15889.14 A15889.22
3-Bromo-o-xylene ≥99%
3-Bromo-o-xylene
Purity: 99% CAS No.: 576-23-8 Molecular Formula: C8H9Br Formula Weight: 185.07 Flash Point: 80°(176°F) Refractive Index: 1.5600 Boiling Point: 212-214° Beilstein Reference: 2077312 MDL No.: MFCD00000069
Formel: C₈H₉Br
Kokpunkt: 212…214 °C
Densitet: 1,365
Flampunkt: 80 °C (176 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000069
CAS nummer: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1,II