Chloro(diphenyl)methane 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Benzhydryl chloride
154660050 154662500 154660500
ACRO154660050EA 500 SEK
ACRO154660050 ACRO154662500 ACRO154660500
Chloro(diphenyl)methane 98%
Chloro(diphenyl)methane
Formel: (C₆H₅)₂CHCl
MW: 202,68 g/mol
Kokpunkt: 270 °C (760 mmHg)
Smältpunkt: 15…17 °C
Densitet: 1,14 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000855
CAS nummer: 90-99-3
EINECS: 202-031-7
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5940 to 1.5970 (20°C, 589 nm)
GC ≥97.5 %