α-Phenylbenzyl bromide ≥90%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Bromodiphenylmethane, α-Phenylbenzyl bromide, Benzhydryl bromide
L02211.14 L02211.22 L02211.36
L02211.14EA 790 SEK
L02211.14 L02211.22 L02211.36
α-Phenylbenzyl bromide ≥90%
α-Phenylbenzyl bromide
Purity: 90+% CAS No.: 776-74-9 Molecular Formula: C13H11Br Formula Weight: 247.14 Flash Point: 110°(230°F) Odour: Pungent Boiling Point: 183-185°/20mm Beilstein Reference: 638544 UN No.: 1770 MDL No.: MFCD00000134
Formel: (C₆H₅)₂CHBr
Kokpunkt: 183…185 °C (20 mmHg)
Smältpunkt: 36…41 °C
Densitet: 1,491 g/cm³ (13 °C)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000134
CAS nummer: 776-74-9
EINECS: 212-279-8
UN: 1770
ADR: 8,II