Toluen, EMPLURA®, extra pure, Supelco®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ200
1.08323.1000 1.08323.2500 1.08323.6025 1.08323.9190 1.08323.9011
1.08323.1000EA 491 SEK
1.08323.1000 1.08323.2500 1.08323.6025 1.08323.9190 1.08323.9011
Toluen, EMPLURA®, extra pure, Supelco®
Toluen
For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues
Formel: C₆H₅CH₃
Kokpunkt: 110,6 °C (1013 hPa)
Smältpunkt: –95 °C
Densitet: 0,867 g/cm³ (20 °C)
Flampunkt: 4 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM