Toluen ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Methylbenzene, Phenylmethane, FM approval
L10967.AU L10967.0F
L10967.AUEA 685 SEK
L10967.AU L10967.0F
Toluen ≥99%
Toluen
Purity: 99% CAS No.: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: 4°(40°F) Refractive Index: 1.4967 Odour: Aromatic Melting Point: ca -93° Boiling Point: 111° Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512
Formel: C₆H₅CH₃
Kokpunkt: 111 °C
Smältpunkt: ∼–93 °C
Densitet: 0,867
Flampunkt: 4 °C (40 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008512
CAS nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 14,9529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM