Toluen, vattenfri ≥99.8%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Methylbenzene, Phenylmethane, FM approval
41464.AK
41464.K2
41464.K7
41464.AKEA
950
SEK
41464.AK
41464.K2
41464.K7
Toluen, vattenfri ≥99.8%
Toluen
Purity: 99.8%
CAS No.: 108-88-3
Molecular Formula: C6H5CH3
Formula Weight: 92.14
Flash Point: 4°(40°F)
Physical Form: Liquid
Density: 0.867
Refractive Index: 1.4967
Odour: Aromatic
Melting Point: -93°
Boiling Point: 111°
Beilstein Reference: 635760
Merck Reference: 14,9529
UN No.: 1294
MDL No.: MFCD00008512
Formel:
C₆H₅CH₃ Kokpunkt: 111 °C Smältpunkt: –93 °C Densitet: 0,867 Flampunkt: 4 °C (40 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00008512 CAS nummer: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 14,9529 |