4-Butylphenol ≥98%

Leverantör: Thermo Scientific
Fara

Synonyms: 1-Butyl-4-hydroxybenzene

A12812.06 A12812.14 A12812.22
A12812.06EA 1475 SEK
A12812.06 A12812.14 A12812.22
4-Butylphenol ≥98%
4-Butylphenol

Purity: 98% CAS No.: 1638-22-8 Molecular Formula: C10H14O Formula Weight: 150.22 Refractive Index: 1.5170 Melting Point: ca 22° Boiling Point: 245° Beilstein Reference: 2042120 UN No.: 3145 MDL No.: MFCD00041750

Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokpunkt: 245 °C
Smältpunkt: ∼22 °C
Densitet: 0,980
Flampunkt: 121 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041750
CAS nummer: 1638-22-8
EINECS: 216-672-5
UN: 3145
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR