4-Methylanisole 99%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Methylanisole
164660010 164660050 164662500 164660025
ACRO164660010EA 2200 SEK
ACRO164660010 ACRO164660050 ACRO164662500 ACRO164660025
4-Methylanisole 99%
4-Methylanisole
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokpunkt: 174 °C (760 mmHg)
Smältpunkt: –50 °C
Densitet: 0,96 g/cm³
Flampunkt: 53 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008413
CAS nummer: 104-93-8
EINECS: 203-253-7
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥98.5 %