3-Methylanisole ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Methoxy-3-methylbenzene
B21455.18 B21455.30
B21455.18EA 870 SEK
B21455.18 B21455.30
3-Methylanisole ≥99%
3-Methylanisole
Purity: 99% CAS No.: 100-84-5 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 54°(129°F) Refractive Index: 1.5130 Odour: Pleasant Boiling Point: 175-176° Beilstein Reference: 907023 UN No.: 3271 MDL No.: MFCD00008395
Formel: C₈H₁₀O
MW: 122.17 g/mol
Kokpunkt: 175…176 °C
Smältpunkt: –47 °C
Densitet: 0.969
Flampunkt: 54 °C (129 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008395
CAS nummer: 100-84-5
EINECS: 202-893-4
UN: 3271
ADR: 3 III