3-Nitrotoluen 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: m-Nitrotoluen
129040050 129045000
ACRO129040050EA 375 SEK
ACRO129040050 ACRO129045000
3-Nitrotoluen 99%
3-Nitrotoluen
Formel: C₇H₇NO₂
MW: 137.14 g/mol
Kokpunkt: 230…231 °C (760 mmHg)
Smältpunkt: 15…16 °C
Densitet: 1.15 g/cm³
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007265
CAS nummer: 99-08-1
EINECS: 202-728-6
UN: 1664
ADR: 6.1 II
Merck Index: 12,06748
Order Now
Specifikation testresultat
Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %