2-Nitrotoluen 99+%
Leverantör: Thermo Fisher Scientific
Fara
129030010 129030025
ACRO129030010EA 1250 SEK
ACRO129030010 ACRO129030025
2-Nitrotoluen 99+%
2-Nitrotoluen
Formel: C₇H₇NO₂
MW: 137.14 g/mol
Kokpunkt: 222 °C (760 mmHg)
Smältpunkt: –9 °C
Densitet: 1.16 g/cm³
Flampunkt: 95 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007157
CAS nummer: 88-72-2
EINECS: 201-853-3
UN: 1664
ADR: 6.1 II
Merck Index: 12,06748
Order Now
Specifikation testresultat
Appearance Clear yellow to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5445 to 1.5465 (20°C, 589 nm)
GC ≥99.0 %