3-Isopropylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Aminocumene, m-Cumidine
I0433-25ML
I0433-500ML
TCIAI0433-25MLEA
365
SEK
TCIAI0433-25ML
TCIAI0433-500ML
3-Isopropylaniline ≥98.0% (by GC, titration analysis)
3-Isopropylaniline
Formel:
C₉H₁₃N MW: 135,21 g/mol Kokpunkt: 80°C Smältpunkt: –63 °C Densitet: 0,9526 g/cm³ (20 °C) Flampunkt: 102°C Storage Temperature: Omgivande |
MDL Number:
MFCD00092918 CAS nummer: 5369-16-4 EINECS: 202-797-2 ADR: 8,III Merck Index: 13,02647 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...