4-Isopropylaniline
Leverantör: Thermo Scientific
Synonyms:
p-Cumidine, 4-Aminocumene
SB00317EA
SB00317EE
MAYBSB00317EAEA
0
SEK
MAYBSB00317EA
MAYBSB00317EE
4-Isopropylaniline
4-Isopropylaniline
Formel:
C₉H₁₃N MW: 135,21 g/mol Kokpunkt: 226 °C (1013 hPa) Smältpunkt: –63 °C Densitet: 0,9526 g/cm³ (20 °C) Flampunkt: 92 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007900 CAS nummer: 99-88-7 EINECS: 202-797-2 UN: 1760 ADR: 8,III Merck Index: 13,02647 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...