4-Isopropylaniline 99%
Leverantör: Thermo Scientific
Fara
Synonyms: p-Cumidine, 4-Aminocumene
174530500 174530100
ACRO174530500EA 1100 SEK
ACRO174530500 ACRO174530100
4-Isopropylaniline 99%
4-Isopropylaniline
Formel: C₉H₁₃N
MW: 135,21 g/mol
Kokpunkt: 226 °C (1013 hPa)
Smältpunkt: –63 °C
Densitet: 0,989 g/cm³
Flampunkt: 92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007900
CAS nummer: 99-88-7
EINECS: 202-797-2
UN: 2735
ADR: 8,II
Merck Index: 13,02647
Order Now
Specifikation testresultat
Appearance Clear light brown to red-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5440 (20°C, 589 nm)
GC ≥98.5 %