4-tert-Butylaniline 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-(tert-Butyl)aniline
198250050 198250250
ACRO198250050EA 1125 SEK
ACRO198250050 ACRO198250250
4-tert-Butylaniline 99%
4-tert-Butylaniline
Formel: C₁₀H₁₅N
Kokpunkt: 230 °C
Smältpunkt: 15…16 °C
Densitet: 0,93 g/cm³
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007899
CAS nummer: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear orange-red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥98.5 %