α,α,α-Trifluoro-4-iodotoluene
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene
TL00447EA
TL00447EE
MAYBTL00447EAEA
0
SEK
MAYBTL00447EA
MAYBTL00447EE
α,α,α-Trifluoro-4-iodotoluene
α,α,α-Trifluoro-4-iodotoluene
Formel:
C₇H₄F₃I MW: 272,01 g/mol Kokpunkt: 183…185 °C (1013 hPa) Smältpunkt: 17 °C Densitet: 1,851 g/cm³ (26 °C) |
MDL Number:
MFCD00039398 CAS nummer: 455-13-0 EINECS: 207-234-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...